Learn More
StatLab PathFlow PiSmart Thermal Transfer Microbiopsy Cassettes
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
Description
- 1,000/bx, 2 bx/case, 2,000/cs
- Threaded in strips of 40 with lids separate
- PathFlow PiSmart Cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing
- Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer
- The new and improved cassettes now incorporate a smoother print face, resulting in improved print of barcodes and text
- Available in 9 colors
Specifications
Specifications
Color | Blue |
Product Type | Microbiopsy Cassettes |
Made in USA