Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PATL2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548187

Catalog No. PA548187


Only null left
Add to Cart

Description

Description

PATL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

RNA-binding protein that acts as a translational repressor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PATL2
Polyclonal
Unconjugated
PATL2
4930424G05Rik; hPat1a; PAT1 homolog 2; Pat1a; PAT1-like protein 2; Patl2; protein associated with topoisomerase II homolog 2; protein associated with topoisomerase II homolog 2 (yeast); protein PAT1 homolog 2; Protein PAT1 homolog a
Rabbit
Antigen affinity chromatography, Protein A
RUO
197135
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
C9JE40
PATL2
KLH conjugated synthetic peptide between 168-196 amino acids from the Central region of human PATL2.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.