Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wiley Patty's Industrial Hygiene, 5th Edition, Volumes 14
Specifications
Category | Safety and Training Materials |
---|---|
Format | Book |
For Use With (Application) | Reference for the fields of occupational health and toxicology |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-041-299
![]() |
Wiley
9780470074848 |
Patty's Industrial Hygiene, Volume 1, Hazard Recognition | N/A | N/A | |||||
Please call customer service for current pricing.
1-800-766-7000 |
|||||||||
19-041-305
![]() |
Wiley
9780470074879 |
Patty's Industrial Hygiene, Volume 4, Program Management and Specialty Areas of Practice | N/A | N/A | |||||
Please call customer service for current pricing.
1-800-766-7000 |
|||||||||
19-041-298
![]() |
Wiley
9780470074855 |
Patty's Industrial Hygiene, Volume 2, Evaluation and Control | N/A | N/A | |||||
Please call customer service for current pricing.
1-800-766-7000 |
|||||||||
19-041-306
![]() |
Wiley
9780470074862 |
Patty's Industrial Hygiene, Volume 3, Physical and Biological Agents | N/A | N/A | |||||
Please call customer service for current pricing.
1-800-766-7000 |
|||||||||
Specifications
Safety and Training Materials | |
Book | |
Reference for the fields of occupational health and toxicology |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title