Patty's Industrial Hygiene, 5th Edition, Volumes 1–4

Manufacturer: John Wiley and Sons 9780470074855

 View more versions of this product

Catalog No. 19-041-298

  Check Availability
Add to Cart

    Specifications

    Product Type Patty's Industrial Hygiene, Volume 2, Evaluation and Control
    Category Safety and Training Materials
    For Use With (Application) Reference for the fields of occupational health and toxicology
    Format Book

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit