Learn More
Novus Biologicals™ PATZ Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP233506PEP
Catalog No. NBP23506PEP
Add to cart
Description
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PATZ1. The PATZ Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Antibody Competition
23598
>80% by SDS-PAGE and Coomassie blue staining
E. coli
RUO
PATZ
PBS and 1M Urea, pH 7.4.
27 kDa
100 μl
Store at -20°C. Avoid freeze-thaw cycles.
PATZ1
Human