Learn More
Paulson A-TAC™ Firefighter Goggles: Wildland
Designed for the wildland firefighter
Manufacturer:  Paulson 510WEB
Catalog No. 19-821-756
Add to cart
Description
Description
  • Polycarbonate, hard-coated/anti-fog lens
  • Fit comfortably over prescription, sun, and safety glasses
  • Indirect ventilation ports
  • Can be placed on the helmet or worn on the head
  • Features the “Quick Strap”
  • Elastic strap with quick release buckle
  • Strap tension adjusts while wearing gloves
Compliance
Meets ANSI Z87.1 standard for impact, penetration and chemical splash exposure
Specifications
Specifications
Firefighter Goggles
Anti-fog
elastic Quick Strap
Baffled top/bottom vents
Clear
ANSI Z87.1
Polycarbonate
Antifog; Hardcoat