Paulson™ Sun Shield for MSA™ V-Guard Hat

Manufacturer: Paulson MfgSupplier Diversity Partner AS4M

 View more versions of this product

Catalog No. 19-816-416

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance OSHA 29 CFR 1910.132 (a), DIN EN 166:2002, DIN EN 172mAnnex II of the PPE-Directive 89/686/EEC; designed to meet ANSI Z87.1
    Faceshield Material Polycarbonate
    For Use With (Application) For additional UV protection during outdoor construction, roadwork, utilities
    Includes Smoke transparent front section for upward vision
    Color Smoke/White
    Material ABS/Lexan
    Product Type Full brim hard hat sun shield
    Weight (English) 3 oz.
    Weight (Metric) 85.04g
    For Use With (Equipment) MSA V-Gard full brim style hardhats

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit