Learn More
PAWR Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP310923100UL
 View more versions of this product
Catalog No. NB126745
Add to cart
Description
Description
PAWR Polyclonal antibody specifically detects PAWR in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
PAWR
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Western Blot, Immunohistochemistry
Western Blot 1.0 ug/ml, Immunohistochemistry
Purified
The immunogen is a synthetic peptide directed towards the middle region of human PAWR (NP_002574). Peptide sequence VNIPAAECLDEYEDDEAGQKERKREDAITQQNTIQNEAVNLLDPGSSYLL
100 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal