Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Pax-5, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Pax-5, Rabbit Polyclonal Antibody.

$172.00 - $1064.00

Specifications

Antigen PAX-5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RB9406P
View Documents
Thermo Scientific™
RB9406P
Purified Ab with BSA and Azide 1mL Each for $1,064.00
Add to cart
 
RB9406P0
View Documents
Thermo Scientific™
RB9406P0
Purified Ab with BSA and Azide 100μL Each for $172.00
Add to cart
 
RB9406P1
View Documents
Thermo Scientific™
RB9406P1
Purified Ab with BSA and Azide 500μL Each for $608.00
Add to cart
 
RB9406R7
View Documents
Thermo Scientific™
RB9406R7
Ready-to-use for IHC (UVLP) 7mL Each for $401.00
Add to cart
 
RB9406R1
View Documents
Thermo Scientific™
RB9406R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Pax-5, Rabbit Polyclonal Antibody.

PAX-5 is a B cell specific activator protein (BSAP). In the early stages of B cell developmemt, PAX-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. PAX-5 is expressed primarily inpro-, pre-, and mature B cells, but not in plasma cells. Interestingly, PAX-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. This indicates that PAX-5 is important in B cell development, but may also have a role in proper neuronal development.

Host Species: Rabbit

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human PAX-5 protein

Molecular Weight: 50kDa

Positive Control: Tonsil, Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

PAX-5
Polyclonal
Rabbit
B-Cell
Synthetic peptide derived from C-terminus of human Pax-5 protein
Western Blot
Unconjugated
IVD
Primary
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit