Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | PAX1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB125133
|
Novus Biologicals
NBP310116100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
PAX1 Polyclonal specifically detects PAX1 in Mouse samples. It is validated for Western Blot.Specifications
PAX1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
paired box 1, paired domain gene HuP48 | |
The immunogen is a synthetic peptide directed towards the middle region of mouse PAX1 (NP_032806.2). Peptide sequence SLAQPGPYEASKQPPPQPALPYNHIYQYPYPSPVSPTGTKMGTHPGVPGS | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
5075 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title