Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax2, Mouse anti-Human, Alexa Fluor 750, Clone: PAX2/1104, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279877AF750
Description
Pax2 Monoclonal specifically detects Pax2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pax2 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PAX2 | |
Recombinant fragment (around aa 223-354) human Pax2 protein (exact sequence is proprietary) (Uniprot: Q02962) | |
0.1 mL | |
Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
This antibody recognizes a protein of 42kDa, which is identified as Pax2. Pax2 has been used as a marker for the identification of renal cell carcinoma and ovarian carcinoma by immunohistochemistry. | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PAX2/1104 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title