Learn More
Abnova™ PAX3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00005077T02
Catalog No. 89-955-773
Add to cart
Specifications
Specifications
293T
NM_181458.1
pCMV-PAX3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
PAX3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 53.5kDa
Transient overexpression cell lysate was tested with anti-PAX3 antibody (Mfr. No. H00005077-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human