Learn More
Pax3 Mouse anti-Human, Mouse, DyLight 350, Clone: 7D8G7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237386UV
Catalog No. NBP237386UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Pax3
7D8G7
Western Blot, ELISA
Purified
PAX3
Purified recombinant fragment of human PAX3 (AA: 142-203) expressed in E. Coli.
Protein A or G purified
Apoptosis, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary
5077
ELISA, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse