Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax5/BSAP Mouse anti-Human, Clone: PAX5/2595, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Pax5/BSAP |
---|---|
Clone | PAX5/2595 |
Concentration | 0.2 mg/ml |
Dilution | ELISA 1-5 ug/ml |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122634
|
Novus Biologicals
NBP30758020UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Pax5/BSAP Monoclonal specifically detects Pax5/BSAP in Human samples. It is validated for ELISA.Specifications
Pax5/BSAP | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
5079 | |
IgG1 κ | |
Protein A or G purified |
PAX5/2595 | |
ELISA 1-5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
Recombinant full-length human Pax5/BSAP protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title