Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax5/BSAP Mouse anti-Human, Clone: PAX5/3735, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Pax5/BSAP
Clone PAX5/3735
Concentration 0.2 mg/ml
Dilution Western Blot, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122636
View Documents
Novus Biologicals
NBP30758120UG
20 μg
Each for $138.37
Add to cart
 
NB122635
View Documents
Novus Biologicals
NBP30758110UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Pax5/BSAP
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
A recombinant fragment (around aa 235-382) of human Pax5/BSAP protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
PAX5/3735
Western Blot, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators
10 mM PBS with 0.05% BSA
5079
IgG1 Lambda
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit