Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax6 Mouse anti-Human, Mouse, Clone: AD2.38, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen Pax6
Clone AD2.38
Concentration 1.0 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5552I100
View Documents
MilliporeSigma
MAB5552I100UG
100 μg
Each for $373.00
Add to cart
 
MAB5552I25
View Documents
MilliporeSigma
MAB5552I25UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

Pax6 Monoclonal antibody specifically detects Pax6 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

Pax6
1.0 mg/mL
Unconjugated
Mouse
Human, Mouse
Recombinant human Pax6.
Monoclonal
Protein G purified
AD2.38
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit