Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX7 Mouse anti-Human, Clone: PAX7/1187, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14392

Catalog No. 89-034-474


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human PAX7.

  • High specificity
  • High reactivity
Specifications

Specifications

PAX7
PAX7/1187
Mouse monoclonal antibody raised against partial recombinant human PAX7.
In 10 mM PBS.
Liquid
PAX7
Recombinant protein corresponding to amino acids 300-600 of human PAX7.
Protein A/G purification
RUO
Primary
5081
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P23759
HUP1|PAX7B
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)