Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax7 Rabbit anti-Human, Alexa Fluor 647, Clone: PAX7/7079R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314034AF647
This item is not returnable.
View return policy
Description
Pax7 Monoclonal antibody specifically detects Pax7 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Pax7 | |
Monoclonal | |
Alexa Fluor 647 | |
FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2 | |
Recombinant fragment (aa301-505) of human Pax7 protein (exact sequence is proprietary) (Uniprot: P23759) | |
0.1 mL | |
Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
5081 | |
Store at 4°C in the dark. | |
IgG κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PAX7/7079R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title