Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
PAX7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205701AP
Catalog No. 50-173-2969
Description
Description
The antibody is specific to PAX7. It will not cross react with other PAX members.
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
PAX7
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23759, P47239
HUP1, paired box 7, Paired box protein Pax 7, PAX7, PAX7B
Rabbit
IgG
150 μL
-20°C
Primary
18509, 5081
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PAX7
Liquid
PAX7
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse