Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX8 Mouse anti-Human, Allophycocyanin/Cy7, Clone: PAX8/1491, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254486APCCY7

Catalog No. N254486APC7


Add to cart

Description

Description

PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

PAX8
PAX8/1491
Flow Cytometry
Conjugated, Purified
PAX8
Recombinant human PAX8 fragment (aa60-261) (exact sequence is proprietary)
Protein A or G purified
Cancer
Primary
7849.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit