Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX8 Rabbit anti-Human, Clone: JJ08-88, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267876
Description
PAX8 Monoclonal specifically detects PAX8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunocytochemistry.Specifications
PAX8 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:500, Immunocytochemistry | |
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
Rabbit | |
Protein A purified | |
Cancer | |
7849 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
JJ08-88 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
PAX8 | |
Recombinant protein within Human PAX8 aa 101-450/ 450. (SwissProt: Q06710 Human) | |
100 μL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction