Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX8 Rabbit anti-Human, Clone: PAX8/2774R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen PAX8
Clone PAX8/2774R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122186
View Documents
Novus Biologicals
NBP30735620UG
20 μg
Each for $159.82
Add to cart
 
NB122185
View Documents
Novus Biologicals
NBP30735610UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

PAX8
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8
Recombinant fragment (around aa 60-261) of human PAX8 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
PAX8/2774R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer
10 mM PBS with 0.05% BSA
7849
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit