Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAX8 Rabbit anti-Human, Clone: PAX8/3688R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | PAX8 |
---|---|
Clone | PAX8/3688R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122194
|
Novus Biologicals
NBP30736020UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
PAX8 Monoclonal specifically detects PAX8 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
PAX8 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer | |
10 mM PBS with 0.05% BSA | |
7849 | |
IgG | |
Protein A or G purified |
PAX8/3688R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
Synthetic peptide corresponding to the C-terminus of human PAX8 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title