Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX8 Rabbit anti-Human, Clone: SD0832, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP267793

Catalog No. NBP267793


Add to Cart

Description

Description

PAX8 Monoclonal specifically detects PAX8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PAX8
Monoclonal
1 mg/mL
Western Blot, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8
Rabbit
Protein A purified
Cancer
7849
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
SD0832
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
PAX8
Recombinant protein within Human PAX8 aa 260-450/ 450. (SwissProt: Q06710 Human; SwissProt: Q00288 Mouse; SwissProt: P51974 Rat)
100 μL
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit