Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX9 Rabbit anti-Human, Clone: SD082-02, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267790

Catalog No. NBP267790


Add to cart

Description

Description

PAX9 Monoclonal antibody specifically detects PAX9 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PAX9
SD082-02
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
paired box 9, paired box gene 9, paired box protein Pax-9, paired domain gene 9, STHAG3
Rabbit
IgG
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
Western Blot 1:1000-1:2000
Purified
PAX9
Recombinant Protein
Protein A purified
Cancer
Primary
5083
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit