Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paxillin Chimeric Recombinant Rabbit Monoclonal Antibody (LD2 sAB), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA547944

Encompass

Catalog No. PIMA547944


Add to Cart

Description

Description

Specificity: The antibody binds specifically to the LD-2 motif of human paxillin with a Kd of 24 nM.

Paxillin Chimeric Recombinant Monoclonal antibody specifically detects Paxillin Chimeric in Human samples. It is validated for ELISA, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Paxillin Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P49023
Rabbit
Affinity chromatography
RUO
5829
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
ELISA, Western Blot
LD2 sAB
Unconjugated
PXN
adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12
Peptide corresponding to human paxillin-LD4 motif (residues 260-278) fused to MBP; peptide sequence: WGGSATRELDELMASLSD.
200 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit