Learn More
Selleck Chemical LLCSupplier Diversity Partner Pazopanib HCl (GW786034 HCl) 25mg 635702-64-6  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S103525MG
Pazopanib HCl (GW786034 HCl) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFR, FGFR, c-Kit and c-Fms with IC50 of 10 nM, 30 nM, 47 nM, 84 nM, 74 nM, 140 nM and 146 nM in cell-free assays, respectively. Pazopanib induces autophagic Type II cell death.
Catalog No. 50-621-2