Learn More
BiolegendSupplier Diversity Partner Pacific Blue™ anti-MU/HU GL7 Antigen (T/B Cell Act. Marker)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 144614
Pacific Blue™ anti-MU/HU GL7 Antigen (T/B Cell Act. Marker) [GL7]; Isotype: Rat IgM, κ; Reactivity: Mouse, Human; Apps: FC; Size: 100 ug
Catalog No. 50-711-899