Learn More
Charles River Labs Cell Solutions Peripheral Blood CD3+ Pan T Cells (Negative Selection), 50 Million cells, Cryopreserved
Supplier:  Charles River Labs Cell Solutions PB03NC4
Encompass_Preferred
CD3+ T cells are isolated from leukopaks by either positive or negative selection (NS) using immunomagnetic cell separation procedures.
Catalog No. 50-225-6290
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.