Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBEF/Visfatin/NAMPT, Mouse anti-Human,Mouse,Rat, Clone: OMNI379, Novus Biologicals™

Mouse Monoclonal Antibody

$261.50 - $450.50

Specifications

Antigen PBEF/Visfatin/NAMPT
Clone OMNI379
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Host Species Mouse
Target Species Human, Mouse, Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013149
View Documents
Novus Biologicals
NBP280036100UG
100 ug
Each for $450.50
Add to cart
 
NB013150
View Documents
Novus Biologicals
NBP28003650UG
50 ug
Each for $261.50
Add to cart
 
Description

Description

PBEF/Visfatin/NAMPT Monoclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human, Mouse, Rat Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

PBEF/Visfatin/NAMPT
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Human, Mouse, Rat
Primary
OMNI379
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit