Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PBK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578536
Description
Positive Control: 293T, A431, HeLa, HepG2, U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (90%), Rat (93%), Pig (93%), Chimpanzee (99%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PBK (TOPK) is a serine/threonine kinase that is related to the dual specific mitogen-activated protein kinase (MAPKK). The kinase is up-regulated in a variety of hematological malignancies, participates in activating lymphoid cells, and supports testicular function (spermatogenesis). Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.
Specifications
PBK | |
Polyclonal | |
Unconjugated | |
PBK | |
2810434B10Rik; AW538537; Cancer/testis antigen 84; CT84; D14Ertd732e; epididymis luminal protein 164; HEL164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PBK; PDZ binding kinase; PDZ-binding kinase; serine/threonine protein kinase; spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase; TOPK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55872 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q96KB5 | |
PBK | |
Recombinant protein encompassing a sequence within the center of human PBK. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction