Learn More
Pbp2B Rabbit anti-Bacillus species, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma ABS219925UL
 View more versions of this product
Catalog No. ABS219925MI
Add to cart
Description
Description
Pbp2B Polyclonal antibody specifically detects Pbp2B in Bacillus species samples. It is validated for Immunofluorescence, Western Blotting.
Specifications
Specifications
Pbp2B
Unconjugated
Unpurified
PbpB
His-tagged full-length recombinant Penicillin-binding protein 2B from Bacillus subtilis, strain 168CA.
∼80 kDa observed; 79.32 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Stable for 1 year at -20°C from date of receipt.
Polyclonal
Bacillus species
Immunofluorescence, Western Blot
NP_389399
Penicillin-binding protein 2B;PBP-2B;Penicillin-binding protein B
Rabbit
IgG
Unpurified
Signaling
Primary
This rabbit polyclonal antibody specifically detects Penicillin-binding protein 2B (Pbp2B) in Bacillus subtilis.