Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBS (Phosphate-buffered Saline), 10X, pH 7.4, Alfa Aesar

Non-toxic to most cells and is used in biological research.

$36.50 - $118.00

Specifications

Concentration 10X
Format Liquid
Color Colorless
pH 7.4
Product Type Phosphate-buffered Saline
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ62036K2 View Documents Alfa Aesar
J62036K2
1L Each for $36.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ62036K3 View Documents Alfa Aesar
J62036K3
2L Each for $63.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ62036K7 View Documents Alfa Aesar
J62036K7
4L Each for $118.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

10X
Colorless
Phosphate-buffered Saline
Liquid
7.4
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit