Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBX, long form Mouse anti-Human, Clone: 359.19, Active Motif™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$116.85 - $356.95

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen PBX, long form
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3189 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61152
10 μg
Each for $116.85
Add to cart
 
50-199-3188 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61151
100 μg
Each for $356.95
Add to cart
 
Description

Description

This gene encodes a nuclear protein that belongs to the PBX homeobox family of transcriptional factors. Studies in mice suggest that this gene may be involved in the regulation of osteogenesis, and required for skeletal patterning and programming. A chromosomal translocation, t(1; 19) involving this gene and TCF3/E2A gene, is associated with pre-B-cell acute lymphoblastic leukemia. The resulting fusion protein, in which the DNA binding domain of E2A is replaced by the DNA binding domain of this protein, transforms cells by constitutively activating transcription of genes regulated by the PBX protein family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Monoclonal
Primary
PBX, long form
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit