Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBX1 Mouse anti-Human, Alexa Fluor 594, Clone: PCRP-PBX1-3C8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314035AF594
This item is not returnable.
View return policy
Description
PBX1 Monoclonal antibody specifically detects PBX1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
PBX1 | |
Monoclonal | |
Alexa Fluor 594 | |
DKFZp686B09108, Homeobox protein PBX1, Homeobox protein PRL, MGC126627, Pre-B cell leukemia transcription factor-1, pre-B-cell leukemia homeobox 1, pre-B-cell leukemia transcription factor 1, PRL | |
Recombinant full-length human PBX1 protein (Uniprot: P40424) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-PBX1-3C8 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
5087 | |
Store at 4°C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title