Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBX1b Monoclonal Antibody (CT 41.1), Active Motif™
SDP

Mouse Monoclonal Antibody

$132.55 - $397.65

Specifications

Antigen PBX1b
Clone CT 41.1
Concentration 1 mg/mL
Applications ChIP Assay, Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3201
SDP
Encompass
View Documents
Active Motif
61166
10 μg
Each for $132.55
Only null left
Add to Cart
 
50-199-3200
SDP
Encompass
View Documents
Active Motif
61165
100 μg
Each for $397.65
Only null left
Add to Cart
 
Description

Description

This gene encodes a nuclear protein that belongs to the PBX homeobox family of transcriptional factors. Studies in mice suggest that this gene may be involved in the regulation of osteogenesis, and required for skeletal patterning and programming. A chromosomal translocation, t(1; 19) involving this gene and TCF3/E2A gene, is associated with pre-B-cell acute lymphoblastic leukemia. The resulting fusion protein, in which the DNA binding domain of E2A is replaced by the DNA binding domain of this protein, transforms cells by constitutively activating transcription of genes regulated by the PBX protein family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

PBX1b
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
2310056B04Rik; D230003C07Rik; DKFZp686B09108; Homeobox protein PBX1; Homeobox protein PRL; MGC126627; OTTHUMP00000032205; PBX homeobox 1; Pbx1; Pbx-1; pre B cell leukemia homeobox 1; pre-B-cell leukemia homeobox 1; Pre-B-cell leukemia transcription factor 1; PRL
PBX1
IgG1 κ
Protein G
Antibody
CT 41.1
ChIP Assay, Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human
P40424
5087
This PBX1b antibody was raised against a recombinant protein corresponding to amino acids 319 - 347 of human PBX1b.
Primary
-20°C, Avoid Freeze/Thaw Cycles
PBX1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.