Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBX2 Mouse anti-Human, Clone: 2.1, Active Motif™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$116.85 - $356.95

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen PBX2
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3193 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61156
10 μg
Each for $116.85
Add to cart
 
50-199-3192 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61155
100 μg
Each for $356.95
Add to cart
 
Description

Description

PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.
Specifications

Specifications

Monoclonal
Primary
PBX2
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit