Learn More
PBX2 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP
Mouse Recombinant Monoclonal Antibody
Supplier:  Active Motif 91227
Encompass
 View more versions of this product
Catalog No. 50-199-3757
Add to Cart
Description
Description
PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.
Specifications
Specifications
PBX2
Monoclonal
Unconjugated
PBX2
AU043397; BOS_21660; G17; homeobox 12; Homeobox protein PBX2; HOX12; LOC100359211; PBX homeobox 2; Pbx2; PBX2MHC; PBX2P1; pbxy; pbxy homeodomain protein; pre B cell leukemia homeobox 2; pre B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia homeobox 2 pseudogene 1; pre-B-cell leukemia homeobox 3; Pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia transcription factor y; Protein G17; wu:fb73g03; XXbac-BPG300A18.13
Mouse
Protein A
RUO
18515, 5089
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5
O35984, P40425
PBX2
This antibody was raised against a recombinant protein corresponding to amino acids 308-430 of mouse PBX2 (pre-B-cell leukemia homeobox 2).
100 μg
Primary
Human, Mouse
Antibody
IgG2a