Learn More
PBX2 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
$96.96 - $442.38
Specifications
Antigen | PBX2 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3758
![]() ![]() |
Active Motif
91228 |
10 μg |
Each for $96.96
|
|
|||||
50-199-3757
![]() ![]() |
Active Motif
91227 |
100 μg |
Each for $442.38
|
|
|||||
Description
PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.Specifications
PBX2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O35984, P40425 | |
18515, 5089 | |
This antibody was raised against a recombinant protein corresponding to amino acids 308-430 of mouse PBX2 (pre-B-cell leukemia homeobox 2). | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
PBX2 |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
AU043397; BOS_21660; G17; homeobox 12; Homeobox protein PBX2; HOX12; LOC100359211; PBX homeobox 2; Pbx2; PBX2MHC; PBX2P1; pbxy; pbxy homeodomain protein; pre B cell leukemia homeobox 2; pre B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia homeobox 2 pseudogene 1; pre-B-cell leukemia homeobox 3; Pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia transcription factor y; Protein G17; wu:fb73g03; XXbac-BPG300A18.13 | |
PBX2 | |
IgG2a | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.