Learn More
Invitrogen™ PBXIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597157
Description
Immunogen sequence: MASCPDSDNS WVLAGSESLP VETLGPASRM DPESERALQA PHSPSKTDGK ELAGTMDGEG TLFQTESPQS GSILTEETEV KGTLEGDVCG VEPPGPGDTV VQGDLQETTV VTGLGPDTQD LEGQSPPQSL PSTPKAAWIR EEGRCSSSDD DTDVDMEGLR RRRGREAGPP QPMVPLAVEN QAGGEGAGGE; Positive Samples: HeLa, LO2, BT-474, A-549; Cellular Location: Cytoplasm, Nucleus, cytoskeleton.
Regulator of pre-B-cell leukemia transcription factors (BPXs) function. Inhibits the binding of PBX1-HOX complex to DNA and blocks the transcriptional activity of E2A-PBX1. Tethers estrogen receptor-alpha (ESR1) to microtubules and allows them to influence estrogen receptors-alpha signaling.
Specifications
PBXIP1 | |
Polyclonal | |
Unconjugated | |
PBXIP1 | |
4732463H20Rik; hematopoietic PBX-interacting protein; HPIP; PBX homeobox interacting protein 1; PBXIP1; pre B cell leukemia transcription factor interacting protein 1; pre-B-cell leukemia homeobox interacting protein 1; pre-B-cell leukemia transcription factor interacting protein 1; pre-B-cell leukemia transcription factor-interacting protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
229534, 310644, 57326 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2VD12, Q3TVI8, Q96AQ6 | |
PBXIP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human PBXIP1 (NP_0653852). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.