Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PC4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124480
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124480 100 μg
1 options

Catalog No. PIPA124480

Supplier: Thermo Scientific PA124480

Rabbit Polyclonal Antibody

PA1-24480 detects PC4 in Human samples. PA1-24480 has been successfully used in Western Blot procedures. PA1-24480 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human PC4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PC4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene SUB1
Gene Accession No. P53999, P11031, Q63396
Gene Alias PC4, TIS7
Gene Symbols LOC100686657, SUB1, SUB1A
Host Species Rabbit
Immunogen Synthetic peptide region of human PC4.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100686657, 10923, 192269, 20024, 479283, 553561
Target Species Canine, Fish, Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.