Learn More
Avon Protection PC50 Enforcer Kit
Based off the widely deployed 50 Series masks and is designed and approved as a NON-CBRN respiratory device, providing protection in dangerous and contaminated environments, against certain solid and liquid respirable particles, gases, vapors and splashes of liquid chemicals.
Manufacturer:  Avon Protection 705016282
 View more versions of this product
Catalog No. 18-000-118
Add to cart
Description
Description
  • Developed for the targeted user groups; budget challenged Prisons, Correctional Officers, Riot Control, Border Patrol, Plant Security and other NON-CBRN requirements that demand a robust, field proven competitively priced military pedigree product that meets their operational needs.
  • Available in three sizes, with either left or right hand 40 mm canister mounts that accept traditional or conformal filters.
  • The standard communication port allows the ability to integrate with existing radios.
  • The PC50 may be fitted with a vision correction system and a variety of outserts for specific operational situations.
  • This product is designed for the lowest cost of ownership and maximum reliability.
  • Kit Includes: PC50 Twin Port Mask, Clear Outsert Assembly, Sunlight Outsert Assembly and CTCF50 Riot Agent Canister
Specifications
Specifications
Yes
Yes
Black
Medium
No
Yes
Class 1
Full Face Respirator
Yes