Learn More
PCB standard Solution, 7, 10 μg/mL in isooctane, MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 3698910ML
 View more versions of this product
Catalog No. 11-101-6493
Add to cart
Description
Description
Environmental
Specifications
Specifications
10 µg/mL in isooctane
PCB No 28, 52, 101, 138, 153, 180, 209 solution
10 mL
Analytical Standard
540-84-1
Analytical Standard
4°C - Closed Cup
Safety and Handling
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label