Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCCB Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297682F
Description
PCCB Polyclonal specifically detects PCCB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCCB | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
beta polypeptide, mitochondrial, propionyl CoA carboxylase, beta polypeptide | |
Produced in rabbits immunized with E. coli-derived Human PCCB fragment. | |
0.1 mL | |
Stem Cell Markers | |
5096 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title