Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCCB Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen PCCB
Applications Immunofluorescence, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093662
View Documents
Novus Biologicals
NBP303620100UL
100 μg
Each for $320.71
Add to cart
 
NB093663
View Documents
Novus Biologicals
NBP30362020UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

PCCB Polyclonal antibody specifically detects PCCB in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence, KnockDown
Specifications

Specifications

PCCB
Unconjugated
Rabbit
Stem Cell Markers
PBS with 50% glycerol, pH7.3.
5096
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, KnockDown, Western Blot
Purified
RUO
Human, Mouse
beta polypeptide, mitochondrial, propionyl CoA carboxylase, beta polypeptide
Recombinant fusion protein containing a sequence corresponding to amino acids 260-539 of human PCCB (NP_000523.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit