Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH11 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179198
SDP
View Documents
Novus Biologicals
NBP179198
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

PCDH11 Polyclonal specifically detects PCDH11 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Western Blot
Unconjugated
RUO
27328
IgG
Polyclonal
Rabbit
NP_001074854
The immunogen for this antibody is Pcdh11x. Peptide sequence LNQSSMLLIKVKDENDNAPVFTQSFISLSVPENNSPGAQLTKISATDADS.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.