Learn More
PCDH11X Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 200701AP
Catalog No. 50-173-2681
Add to cart
Description
Description
The antibody recognizes isoforms 1 and isoform 6 of PCDH11X. The antibody has no cross reaction to PCDH11Y.
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its Y homolog, despite divergence leading to coding region changes, is the most closely related cadherin family member. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Disruption of this gene may be associated with developmental dyslexia. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
PCDH11X
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BZA7
KIAA1326, PCDH X, PCDH11, PCDH11X, PCDHX, PCDHY, Protocadherin 11, protocadherin 11 X linked, Protocadherin S
Rabbit
IgG
150 μL
-20°C
Primary
27328
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PCDH11X
Liquid
PCDH11X
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human