Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH11X Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17919820UL

 View more versions of this product

Catalog No. NBP17919820


Add to cart

Description

Description

PCDH11X Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

PCDH11X
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
PCDH11X
The immunogen for this antibody is Pcdh11x. Peptide sequence LNQSSMLLIKVKDENDNAPVFTQSFISLSVPENNSPGAQLTKISATDADS.
Immunogen affinity purified
RUO
Primary
27328
Western Blot
Western Blot 1:1000
NP_001074854
KIAA1326, PCDH11protocadherin-S, PCDHXprotocadherin 11X, PCDH-Xprotocadherin-11 X-linked, protocadherin 11, protocadherin 11 X-linked, Protocadherin on the X chromosome, Protocadherin-11, Protocadherin-S
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit