Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDH9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548525

Catalog No. PA548525


Add to cart

Description

Description

Potential calcium-dependent cell-adhesion protein.
Specifications

Specifications

PCDH9
Polyclonal
PBS with 0.09% sodium azide
Q9HC56
Protocadherin-9, PCDH9
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5101
Western Blot
Unconjugated
PCDH9
Liquid
PCDH9
KLH conjugated synthetic peptide between 1018-1046 amino acids from the C-terminal region of human PCDH9.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit