Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHA6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PCDHA6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15926220
![]() |
Novus Biologicals
NBP15926220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159262
![]() |
Novus Biologicals
NBP159262 |
100 μL |
Each for $487.50
|
|
|||||
Description
PCDHA6 Polyclonal specifically detects PCDHA6 in Human samples. It is validated for Western Blot.Specifications
PCDHA6 | |
Polyclonal | |
Rabbit | |
Q9UN73-3 | |
56142 | |
Synthetic peptides corresponding to PCDHA6(protocadherin alpha 6) The peptide sequence was selected from the C terminal of PCDHA6. Peptide sequence LVKDHGEPALTATATVLVSLVESGQAPKASSRASVGAAGPEAALVDVNVY. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CNR2, CNRN2, CNRS2, CRNR2, KIAA0345-like 8, PCDH-alpha-6, PCDH-ALPHA6, protocadherin alpha 6, protocadherin alpha-6 | |
PCDHA6 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title